Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTSS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138383.2 | NP_612392.1 | 15 | 2101 | ||
XM_005256236.4 | XP_005256293.2 | 14 | 2152 | |||
XM_011523450.2 | XP_011521752.1 | 17 | 2062 | |||
XM_011523453.2 | XP_011521755.2 | 16 | 2314 | |||
XM_011523454.2 | XP_011521756.2 | 15 | 2305 | |||
XM_011523458.2 | XP_011521760.1 | 10 | 1539 | |||
XM_017023879.1 | XP_016879368.1 | 16 | 2218 | |||
XM_017023880.1 | XP_016879369.1 | 14 | 2182 | |||
XM_017023881.1 | XP_016879370.1 | 15 | 2161 | |||
GenBank mRNA | AB115770.1 | - | 15 | 1841 |