Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTSS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138383.2 | NP_612392.1 | 6, 8 | 720 | ||
XM_011523450.2 | XP_011521752.1 | 7, 9 | 624 | |||
XM_011523453.2 | XP_011521755.2 | 6, 8 | 876 | |||
XM_017023879.1 | XP_016879368.1 | 6, 8 | 876 | |||
XM_017023880.1 | XP_016879369.1 | 6, 8 | 876 | |||
XM_017023881.1 | XP_016879370.1 | 6, 8 | 876 | |||
XM_017023882.1 | XP_016879371.1 | 6, 8 | 876 | |||
XM_017023883.1 | XP_016879372.1 | 6, 8 | 876 | |||
GenBank mRNA | AB115770.1 | - | 6, 8 | 460 |