Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138387.3 | NP_612396.1 | 4 | 718 | ||
NR_028581.1 | 5 | 912 | ||||
NR_028582.1 | 4 | 777 | ||||
XM_011525473.2 | XP_011523775.1 | 5 | 472 | |||
XM_011525474.2 | XP_011523776.1 | 5 | 941 | |||
XM_017025335.1 | XP_016880824.1 | 4 | 463 | |||
GenBank mRNA | BC002494.2 | - | 4 | 516 | 3050476 | |
BC021574.1 | - | 4 | 698 | 4842511 |