Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM114A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138389.2 | NP_612398.2 | 10 | 1404 | ||
NR_033290.1 | 8 | 899 | ||||
XM_005262672.2 | XP_005262729.1 | 9 | 1210 | |||
XM_006714033.2 | XP_006714096.1 | 9 | 975 | |||
GenBank mRNA | AK094179.1 | - | 2 | 154 | ||
AK292831.1 | - | 10 | 1331 | |||
AK296859.1 | - | 6 | 586 | |||
AK304050.1 | - | 8 | 881 | |||
AK308959.1 | - | 1 | 219 | |||
BC001096.2 | - | 8 | 1155 | 3507281 | ||
BC040452.1 | - | 10 | 1321 | 3846822 | ||
BX640862.1 | - | 7 | 640 |