Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM114A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138389.2 | NP_612398.2 | 14, 15 | 1859 | ||
NR_033290.1 | 12, 13 | 1354 | ||||
XM_005262672.2 | XP_005262729.1 | 13, 14 | 1665 | |||
XM_006714033.2 | XP_006714096.1 | 13, 14 | 1430 | |||
GenBank mRNA | AK094179.1 | - | 6, 7 | 609 | ||
AK292831.1 | - | 14, 15 | 1786 | |||
AK296859.1 | - | 10, 11 | 1041 | |||
AK304050.1 | - | 12, 13 | 1336 | |||
AK308959.1 | - | 5, 6 | 674 | |||
BC001096.2 | - | 12, 13 | 1610 | 3507281 | ||
BC040452.1 | - | 14, 15 | 1776 | 3846822 | ||
BX640862.1 | - | 11, 12 | 1095 |