Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139072.3 | NP_620711.3 | 12, 13 | 2248 | ||
XM_005246950.2 | XP_005247007.1 | 11, 12 | 2112 | |||
GenBank mRNA | AF442487.1 | - | 12, 13 | 2161 | ||
AK289553.1 | - | 12, 13 | 2206 | |||
AK304631.1 | - | 11, 12 | 1600 | |||
AK308576.1 | - | 8, 9 | 951 | |||
AL137311.1 | - | 5, 6 | 646 | |||
AY358891.1 | - | 12, 13 | 2211 | |||
BC024766.2 | - | 8, 9 | 1254 | |||
BC035009.1 | - | 12, 13 | 2238 | 4820343 |