Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033197.2 | NP_149974.2 | 12 | 1337 | ||
GenBank mRNA | AF364078.1 | - | 12 | 1262 | ||
AK127971.1 | - | 9 | 836 | |||
AK290243.1 | - | 12 | 1319 | |||
AK313176.1 | - | 12 | 1255 | |||
AW449208.1 | - | 5 | 403 | |||
AY358595.1 | - | 12 | 1249 | |||
BC008429.1 | - | 12 | 1252 | 4291561 |