Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145038.4 | NP_659475.2 | 10 | 1446 | ||
XM_005264637.3 | XP_005264694.1 | 9 | 1254 | |||
XM_005264638.3 | XP_005264695.1 | 6 | 801 | |||
XM_017005271.1 | XP_016860760.1 | 9 | 1214 | |||
GenBank mRNA | AK057222.1 | - | 10 | 1442 | ||
AK289953.1 | - | 10 | 1429 | |||
AL833892.1 | - | 9 | 1180 | |||
BC009488.2 | - | 9 | 1169 |