Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145038.4 | NP_659475.2 | 1, 2 | 241 | ||
XM_005264637.3 | XP_005264694.1 | 1, 2 | 233 | |||
XM_017005271.1 | XP_016860760.1 | 1, 2 | 149 | |||
GenBank mRNA | AK057222.1 | - | 1, 2 | 237 | ||
AK289953.1 | - | 1, 2 | 224 | |||
AK307094.1 | - | 1, 2 | 215 | |||
DB021259.1 | - | 1, 2 | 241 |