Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138392.3 | NP_612401.2 | 13 | 1357 | ||
XM_006723474.1 | XP_006723537.1 | 13 | 1387 | |||
XM_006723475.1 | XP_006723538.1 | 13 | 1387 | |||
XM_011527492.1 | XP_011525794.1 | 10 | 1036 | |||
XM_011527493.1 | XP_011525795.1 | 12 | 1283 | |||
XM_017027473.1 | XP_016882962.1 | 13 | 1357 | |||
XM_017027474.1 | XP_016882963.1 | 12 | 1253 | |||
GenBank mRNA | AF258553.1 | - | 13 | 1356 | ||
AK075057.1 | - | 13 | 1279 | |||
AK123695.1 | - | 11 | 1335 | |||
AK297006.1 | - | 7 | 983 | |||
AK300808.1 | - | 11 | 1236 | |||
AK312748.1 | - | 13 | 1321 | |||
AY007117.1 | - | 4 | 392 | |||
BC007653.1 | - | 5 | 643 | |||
BC022855.1 | - | 13 | 1328 | 4102901 |