Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320304.1 | NP_001307233.1 | 7 | 1321 | ||
NM_001320305.1 | NP_001307234.1 | 7 | 1273 | |||
NM_001320306.1 | NP_001307235.1 | 7 | 1340 | |||
NM_001320307.1 | NP_001307236.1 | 7 | 1367 | |||
NM_001320309.1 | NP_001307238.1 | 6 | 1477 | |||
NM_001320310.1 | NP_001307239.1 | 7 | 1354 | |||
NM_001320311.1 | NP_001307240.1 | 7 | 1345 | |||
NM_033416.2 | NP_219484.1 | 7 | 1327 | |||
GenBank mRNA | AF054996.1 | - | 7 | 1142 | ||
AK289878.1 | - | 7 | 655 | |||
AK292345.1 | - | 7 | 655 | |||
AK312808.1 | - | 7 | 655 | |||
AY364245.1 | - | 6 | 619 | |||
BC010042.1 | - | 7 | 636 | 3629513 |