Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142650.1 | NP_001136122.1 | 9 | 1152 | ||
NM_138394.3 | NP_612403.2 | 8 | 1408 | |||
XM_005264639.3 | XP_005264696.2 | 8 | 1405 | |||
XM_005264640.3 | XP_005264697.2 | 9 | 1151 | |||
XM_011533165.2 | XP_011531467.1 | 7 | 1333 | |||
XM_011533166.2 | XP_011531468.1 | 8 | 1405 | |||
GenBank mRNA | AK291462.1 | - | 8 | 1273 | ||
AK292259.1 | - | 9 | 1145 | |||
AL512692.1 | - | 8 | 885 | |||
AY236962.1 | - | 7 | 1135 | |||
AY236963.2 | - | 9 | 997 | |||
BC017480.1 | - | 8 | 1254 | 3502860 | ||
KU178839.1 | - | 8 | 998 | |||
KU178840.1 | - | 8 | 998 | |||
KU178841.1 | - | 9 | 1081 |