Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138395.3 | NP_612404.1 | 1 | 1016 | ||
GenBank mRNA | AB107013.1 | - | 1 | 959 | ||
AK098121.1 | - | 1 | 951 | |||
AK301252.1 | - | 2 | 731 | |||
BC009115.1 | - | 1 | 919 | |||
BC126294.1 | - | 1 | 954 | 8992010 |