Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270539.1 | NP_001257468.1 | 2 | 211 | ||
NM_001300881.1 | NP_001287810.1 | 2 | 176 | |||
NM_080662.3 | NP_542393.1 | 2 | 211 | |||
XM_005272506.2 | XP_005272563.1 | 2 | 213 | |||
XM_011528426.2 | XP_011526728.1 | 2 | 213 | |||
XM_011528427.2 | XP_011526729.1 | 2 | 213 | |||
XM_011528428.1 | XP_011526730.1 | 4 | 470 | |||
XM_011528429.2 | XP_011526731.1 | 4 | 639 | |||
XM_011528431.2 | XP_011526733.1 | 3 | 403 | |||
XM_011528432.1 | XP_011526734.1 | 2 | 213 | |||
GenBank mRNA | AB095921.1 | - | 2 | 201 | ||
AL833945.1 | - | 2 | 176 | |||
BC008780.2 | - | 2 | 192 | 3610362 | ||
BI833221.1 | - | 2 | 144 | |||
HY002496.1 | - | 2 | 212 |