Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138396.5 | NP_612405.2 | 3 | 1123 | ||
GenBank mRNA | AK093897.1 | - | 1 | 38 | ||
AK313925.1 | - | 1 | 800 | |||
BC009489.2 | - | 1 | 603 | 3936171 | ||
BC036455.2 | - | 3 | 1104 | 4811410 | ||
BC050397.1 | - | 3 | 1123 |