Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002006.2 | NP_001002006.1 | 6, 7 | 1181 | ||
NM_001199086.1 | NP_001186015.1 | 6, 7 | 1130 | |||
NM_001199087.1 | NP_001186016.1 | 6, 7 | 1232 | |||
NM_001199088.1 | NP_001186017.1 | 6, 7 | 1187 | |||
NM_033253.3 | NP_150278.2 | 5, 6 | 1001 | |||
GenBank mRNA | AF352326.1 | - | 1, 2 | 139 | ||
AF417165.1 | - | 6, 7 | 1147 | |||
AJ295254.1 | - | 1, 2 | 97 | |||
AK093234.1 | - | 2, 3 | 284 | |||
AK093464.1 | - | 5, 6 | 926 | |||
AK097422.1 | - | 5, 6 | 990 | |||
AK302084.1 | - | 5, 6 | 947 | |||
AK302200.1 | - | 6, 7 | 1076 | |||
AK302795.1 | - | 6, 7 | 1216 | |||
BC136387.1 | - | 5, 6 | 954 | 9020374 | ||
BC171761.1 | - | 5, 6 | 954 | 9054414 |
Gene Symbol: NT5C1B-RDH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199103.1 | NP_001186032.1 | 5, 6 | 1007 | ||
NM_001199104.1 | NP_001186033.1 | 6, 7 | 1181 | |||
GenBank mRNA | AK057444.1 | - | 6, 7 | 1163 | ||
AK307653.1 | - | 5, 6 | 991 |