Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011655.2 | NP_001011655.1 | 6, 7 | 982 | ||
NM_001166305.1 | NP_001159777.1 | 7, 8 | 1123 | |||
NM_001166306.1 | NP_001159778.1 | 6, 7 | 982 | |||
NM_138399.4 | NP_612408.3 | 6, 7 | 982 | |||
XM_005269371.4 | XP_005269428.1 | 6, 7 | 996 | |||
XM_011513318.2 | XP_011511620.1 | 7, 8 | 1026 | |||
XM_011513319.2 | XP_011511621.1 | 6, 7 | 917 | |||
XM_011513320.2 | XP_011511622.1 | 7, 8 | 1147 | |||
XM_011513321.2 | XP_011511623.1 | 6, 7 | 893 | |||
XM_011513322.2 | XP_011511624.1 | 6, 7 | 1003 | |||
XM_017007517.1 | XP_016863006.1 | 6, 7 | 884 | |||
XM_017007518.1 | XP_016863007.1 | 6, 7 | 879 | |||
GenBank mRNA | AL833026.1 | - | 1, 2 | 540 | ||
BC007772.2 | - | 1, 2 | 128 | |||
BC014883.1 | - | 6, 7 | 842 | 3907191 | ||
BC034353.1 | - | 1, 2 | 50 | |||
BC111485.1 | - | 7, 8 | 1082 | 5767916 | ||
BC130297.1 | - | 6, 7 | 886 | 40146328 | ||
BC144158.1 | - | 6, 7 | 886 | 9052678 | ||
BC144159.1 | - | 6, 7 | 886 | 9052679 | ||
BU521517.1 | - | 5, 6 | 538 | |||
BX647418.1 | - | 6, 7 | 741 | |||
CF265142.1 | - | 1, 2 | 137 |