Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORAI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152288.2 | NP_689501.1 | 2 | 716 | ||
GenBank mRNA | AK298276.1 | - | 2 | 678 | ||
BC006126.1 | - | 2 | 659 | 3603998 | ||
BC015555.1 | - | 2 | 705 | 3878011 | ||
BC022786.1 | - | 2 | 659 | 3605904 |