Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145167.2 | NP_660150.1 | 1 | 1049 | ||
GenBank mRNA | AB028127.1 | - | 1 | 990 | ||
AK074655.1 | - | 1 | 1016 | |||
BC001803.1 | - | 1 | 906 | 3357207 | ||
BC019865.1 | - | 1 | 1014 | 4931239 |