Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144577.3 | NP_653178.3 | 8 | 1490 | ||
XM_005259413.3 | XP_005259470.1 | 10 | 1198 | |||
XM_005259414.3 | XP_005259471.1 | 10 | 1194 | |||
XM_005259415.3 | XP_005259472.1 | 10 | 1212 | |||
XM_005259416.4 | XP_005259473.1 | 3 | 345 | |||
XM_011527515.2 | XP_011525817.1 | 11 | 2141 | |||
XM_011527516.2 | XP_011525818.1 | 9 | 931 | |||
XM_011527517.1 | XP_011525819.1 | 10 | 1194 | |||
XM_017027483.1 | XP_016882972.1 | 9 | 1971 | |||
GenBank mRNA | AK057488.1 | - | 2 | 188 | ||
AK128144.1 | - | 10 | 1238 | |||
AL122083.1 | - | 2 | 109 | |||
BC025752.1 | - | 8 | 935 | |||
BC117431.1 | - | 2 | 288 | 40125982 |