Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173470.1 | NP_775741.1 | 4 | 650 | ||
GenBank mRNA | AK295829.1 | - | 5 | 627 | ||
AK312409.1 | - | 4 | 663 | |||
AL157477.1 | - | 4 | 649 | |||
BC033588.1 | - | 4 | 650 | 5213486 |