Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK1IP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144578.3 | NP_653179.1 | 3 | 366 | ||
XM_011537362.2 | XP_011535664.2 | 3 | 1210 | |||
XM_017021789.1 | XP_016877278.1 | 3 | 402 | |||
GenBank mRNA | AK025580.1 | - | 3 | 329 | ||
AK315501.1 | - | 3 | 363 | |||
AL834285.1 | - | 3 | 349 | |||
BC015621.2 | - | 3 | 327 | 4843370 |