Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDR42E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145168.2 | NP_660151.2 | 3 | 984 | ||
XM_005256257.3 | XP_005256314.1 | 4 | 1189 | |||
XM_011523471.2 | XP_011521773.1 | 3 | 998 | |||
XM_011523472.2 | XP_011521774.1 | 3 | 987 | |||
GenBank mRNA | AK026740.1 | - | 3 | 971 | ||
AK315652.1 | - | 3 | 945 | |||
BC019670.1 | - | 3 | 937 | 4517213 |