Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007033.1 | NP_001007034.1 | 4 | 375 | ||
NM_001317999.1 | NP_001304928.1 | 3 | 285 | |||
GenBank mRNA | AY321309.1 | - | 4 | 375 | ||
AY365135.1 | - | 4 | 320 | |||
BC132933.1 | - | 4 | 281 | 40146955 | ||
BC132935.1 | - | 4 | 281 | 40146957 |