Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322977.1 | NP_001309906.1 | 5, 6 | 693 | ||
NM_001322978.1 | NP_001309907.1 | 5, 6 | 636 | |||
NM_001322980.1 | NP_001309909.1 | 5, 6 | 665 | |||
NM_001322981.1 | NP_001309910.1 | 5, 6 | 693 | |||
NM_001322982.1 | NP_001309911.1 | 5, 6 | 664 | |||
NM_001322983.1 | NP_001309912.1 | 5, 6 | 665 | |||
NM_022754.6 | NP_073591.2 | 5, 6 | 665 | |||
GenBank mRNA | AF327346.1 | - | 5, 6 | 575 | ||
AK022287.1 | - | 5, 6 | 727 | |||
AK022938.1 | - | 3, 4 | 299 | |||
AK056915.1 | - | 5, 6 | 570 | |||
BC020517.1 | - | 4, 5 | 460 | 3888861 | ||
BC063241.1 | - | 5, 6 | 583 | 6187994 | ||
BX648188.1 | - | 5, 6 | 581 |