Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322977.1 | NP_001309906.1 | 6 | 739 | ||
NM_001322978.1 | NP_001309907.1 | 6 | 682 | |||
NM_001322980.1 | NP_001309909.1 | 6 | 711 | |||
NM_001322981.1 | NP_001309910.1 | 6 | 739 | |||
NM_001322982.1 | NP_001309911.1 | 6 | 710 | |||
NM_001322983.1 | NP_001309912.1 | 6 | 711 | |||
NM_022754.6 | NP_073591.2 | 6 | 711 | |||
GenBank mRNA | AF327346.1 | - | 6 | 621 | ||
AK022287.1 | - | 6 | 773 | |||
AK022938.1 | - | 4 | 345 | |||
AK056915.1 | - | 6 | 616 | |||
BC020517.1 | - | 5 | 506 | 3888861 | ||
BC063241.1 | - | 6 | 629 | 6187994 | ||
BX648188.1 | - | 6 | 627 |