Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257158.1 | NP_001244087.1 | 2, 3 | 344 | ||
NM_018718.2 | NP_061188.1 | 2, 3 | 344 | |||
NR_046443.1 | 2, 3 | 344 | ||||
GenBank mRNA | AF429308.1 | - | 2, 3 | 190 | ||
AJ278890.1 | - | 2, 3 | 147 | |||
AK026098.1 | - | 2, 3 | 144 | |||
AK311382.1 | - | 2, 3 | 161 | |||
AK314676.1 | - | 2, 3 | 123 | |||
BC018789.1 | - | 2, 3 | 126 | |||
BC056162.1 | - | 2, 3 | 117 | 6142659 | ||
CB155730.1 | - | 2, 3 | 371 | |||
DB069500.1 | - | 2, 3 | 346 |