Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317902.1 | NP_001304831.1 | 3 | 1370 | ||
NM_024831.7 | NP_079107.6 | 4 | 1435 | |||
XM_005251328.2 | XP_005251385.1 | 3 | 1150 | |||
XM_006716485.2 | XP_006716548.1 | 4 | 1221 | |||
XM_017014004.1 | XP_016869493.1 | 4 | 1221 | |||
GenBank mRNA | AF286340.1 | - | 4 | 1525 | ||
AF432215.1 | - | 4 | 1183 | |||
AK055178.1 | - | 1 | 309 | |||
AK314693.1 | - | 4 | 1209 | |||
AY028423.1 | - | 4 | 1219 | |||
AY534911.1 | - | 1 | 225 | |||
BX648548.1 | - | 3 | 1133 |