Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024831.7 | NP_079107.6 | 11 | 2634 | ||
XM_005251328.2 | XP_005251385.1 | 10 | 2349 | |||
GenBank mRNA | AF286340.1 | - | 11 | 2724 | ||
AF432215.1 | - | 11 | 2382 | |||
AK026648.1 | - | 3 | 411 | |||
AK055178.1 | - | 8 | 1508 | |||
AK314693.1 | - | 11 | 2408 | |||
AL832426.1 | - | 5 | 890 | |||
AY028423.1 | - | 11 | 2415 | |||
AY534911.1 | - | 8 | 1424 | |||
BC011999.1 | - | 4 | 528 |