Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPUSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324086.1 | NP_001311015.1 | 2, 3 | 324 | ||
NM_001324410.1 | NP_001311339.1 | 2, 3 | 225 | |||
NM_001324411.1 | NP_001311340.1 | 2, 3 | 324 | |||
NM_001324413.1 | NP_001311342.1 | 2, 3 | 225 | |||
NM_001324414.1 | NP_001311343.1 | 2, 3 | 324 | |||
NM_001324415.1 | NP_001311344.1 | 2, 3 | 324 | |||
NM_058192.2 | NP_478072.1 | 2, 3 | 324 | |||
GenBank mRNA | AL514840.3 | - | 2, 3 | 175 | ||
AL520203.3 | - | 2, 3 | 266 | |||
BC011783.2 | - | 1, 2 | 189 | 3937184 | ||
BG686882.1 | - | 2, 3 | 323 | |||
BM763460.1 | - | 2, 3 | 311 | |||
CN311777.1 | - | 2, 3 | 281 | |||
CN311779.1 | - | 2, 3 | 183 | |||
CX786721.1 | - | 2, 3 | 274 |