Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138423.3 | NP_612432.2 | 6 | 1070 | ||
NM_177974.2 | NP_816929.1 | 6 | 1070 | |||
XM_017021880.1 | XP_016877369.1 | 6 | 1165 | |||
GenBank mRNA | AK095068.1 | - | 2 | 110 | ||
AK291026.1 | - | 6 | 827 | |||
AK298564.1 | - | 7 | 785 | |||
AL832333.1 | - | 6 | 842 | |||
AL832840.1 | - | 6 | 512 | |||
AY358086.1 | - | 6 | 1021 | |||
BC012124.1 | - | 5 | 959 | |||
BC063480.1 | - | 6 | 1024 | 5218924 |