Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138423.3 | NP_612432.2 | 6, 7 | 1128 | ||
NM_177974.2 | NP_816929.1 | 6, 7 | 1128 | |||
GenBank mRNA | AK095068.1 | - | 2, 3 | 168 | ||
AK291026.1 | - | 6, 7 | 885 | |||
AK298564.1 | - | 7, 8 | 843 | |||
AL832333.1 | - | 6, 7 | 900 | |||
AL832840.1 | - | 6, 7 | 570 | |||
AY358086.1 | - | 6, 7 | 1079 | |||
BC063480.1 | - | 6, 7 | 1082 | 5218924 |