Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144582.2 | NP_653183.2 | 6 | 639 | ||
GenBank mRNA | AK056410.1 | - | 2 | 857 | ||
AK074840.1 | - | 4 | 289 | |||
AK303594.1 | - | 6 | 594 | |||
AK315543.1 | - | 6 | 519 | |||
AY358783.1 | - | 6 | 599 | |||
BC010656.1 | - | 4 | 277 | |||
BC020251.1 | - | 4 | 391 | |||
BC128461.1 | - | 6 | 590 | 40126644 | ||
BC128462.1 | - | 6 | 590 | 40126645 | ||
BI552105.1 | - | 6 | 644 |