Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164824.1 | NP_001158296.1 | 3 | 941 | ||
NM_001164825.1 | NP_001158297.1 | 2 | 834 | |||
NM_001320261.1 | NP_001307190.1 | 2 | 716 | |||
NM_138428.5 | NP_612437.3 | 2 | 691 | |||
XM_005270403.4 | XP_005270460.1 | 2 | 1174 | |||
XM_011540553.2 | XP_011538855.1 | 2 | 576 | |||
XM_011540554.2 | XP_011538856.2 | 2 | 1887 | |||
XM_011540555.2 | XP_011538857.1 | 2 | 703 | |||
XM_017000166.1 | XP_016855655.1 | 2 | 695 | |||
GenBank mRNA | AK074853.1 | - | 2 | 623 | ||
BC011880.1 | - | 2 | 617 | 3607345 | ||
BC034598.1 | - | 2 | 2432 | |||
BC064320.1 | - | 2 | 654 | |||
BC073851.1 | - | 2 | 617 | 5499672 | ||
BC093838.1 | - | 1 | 312 | 7939683 | ||
BC107421.1 | - | 2 | 616 | 4688408 | ||
BC112231.1 | - | 1 | 312 | 8327699 | ||
BI759434.1 | - | 2 | 656 |