Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164824.1 | NP_001158296.1 | 3 | 1235 | ||
NM_001164825.1 | NP_001158297.1 | 2 | 1128 | |||
NM_001320261.1 | NP_001307190.1 | 2 | 1010 | |||
NM_138428.5 | NP_612437.3 | 2 | 985 | |||
XM_005270403.4 | XP_005270460.1 | 2 | 1468 | |||
XM_011540553.2 | XP_011538855.1 | 2 | 870 | |||
XM_011540554.2 | XP_011538856.2 | 2 | 2181 | |||
XM_011540555.2 | XP_011538857.1 | 2 | 997 | |||
XM_017000166.1 | XP_016855655.1 | 2 | 989 | |||
GenBank mRNA | AK074853.1 | - | 2 | 917 | ||
BC011880.1 | - | 2 | 911 | 3607345 | ||
BC034598.1 | - | 2 | 2726 | |||
BC064320.1 | - | 2 | 948 | |||
BC073851.1 | - | 2 | 911 | 5499672 | ||
BC093838.1 | - | 1 | 606 | 7939683 | ||
BC107421.1 | - | 2 | 910 | 4688408 | ||
BC112231.1 | - | 1 | 606 | 8327699 |