Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052882.1 | NP_443114.1 | 5 | 778 | ||
GenBank mRNA | AF365931.1 | - | 5 | 778 | ||
BC069071.1 | - | 3 | 393 | 7216885 | ||
BC114446.1 | - | 5 | 577 | 40084041 | ||
BC114503.1 | - | 5 | 577 | 40084040 |