Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135163.1 | NP_001128635.1 | 4, 5 | 878 | ||
NM_052884.2 | NP_443116.2 | 4, 5 | 878 | |||
XM_005258477.3 | XP_005258534.2 | 4, 5 | 819 | |||
GenBank mRNA | AB072038.1 | - | 4, 5 | 797 | ||
AF337818.1 | - | 4, 5 | 878 | |||
AY358135.1 | - | 4, 5 | 756 |