Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017922.1 | NP_001017922.1 | 12 | 1590 | ||
NM_018538.3 | NP_061008.2 | 11 | 1486 | |||
XM_006710313.3 | XP_006710376.1 | 13 | 1855 | |||
XM_011540570.2 | XP_011538872.1 | 12 | 1546 | |||
XM_011540571.2 | XP_011538873.1 | 11 | 1553 | |||
GenBank mRNA | AF311284.1 | - | 1 | 461 | ||
AF311285.1 | - | 11 | 1486 | |||
AK125447.1 | - | 11 | 1334 | |||
AK314436.1 | - | 12 | 1574 | |||
AY049028.1 | - | 11 | 1504 | |||
BC099703.1 | - | 11 | 1419 | 40000626 | ||
BC099707.1 | - | 11 | 1424 | 40000628 | ||
BC099712.1 | - | 11 | 1419 | 40000629 | ||
BC099713.1 | - | 11 | 1419 | 40000630 | ||
BX537371.1 | - | 9 | 2088 |