Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099272.1 | NP_001092742.1 | 3 | 841 | ||
NM_001172418.1 | NP_001165889.1 | 2 | 626 | |||
NM_052893.1 | NP_443125.1 | 4 | 1041 | |||
NM_152733.2 | NP_689946.2 | 2 | 578 | |||
XM_011514279.2 | XP_011512581.1 | 5 | 1438 | |||
XM_011514280.2 | XP_011512582.1 | 3 | 1119 | |||
XM_011514281.2 | XP_011512583.1 | 3 | 1119 | |||
XM_011514282.2 | XP_011512584.1 | 3 | 1119 | |||
XM_017010227.1 | XP_016865716.1 | 4 | 1319 | |||
GenBank mRNA | AB067467.1 | - | 3 | 707 | ||
AK057507.1 | - | 2 | 578 | |||
BC101354.2 | - | 2 | 499 | 40025568 | ||
BC101355.1 | - | 2 | 499 | 40025569 | ||
BC101356.2 | - | 2 | 499 | |||
BC101357.1 | - | 2 | 547 | 40025572 | ||
BX537534.1 | - | 3 | 574 |