Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052897.3 | NP_443129.3 | 11 | 3063 | ||
XM_005268617.1 | XP_005268674.1 | 11 | 3119 | |||
XM_005268618.1 | XP_005268675.1 | 11 | 3215 | |||
XM_005268620.2 | XP_005268677.1 | 11 | 3119 | |||
XM_006719217.1 | XP_006719280.1 | 11 | 2964 | |||
XM_006719218.1 | XP_006719281.1 | 11 | 2952 | |||
XM_011537851.2 | XP_011536153.1 | 11 | 3952 | |||
GenBank mRNA | AB067474.1 | - | 8 | 2732 | ||
AK056399.1 | - | 11 | 3097 | |||
AK093078.1 | - | 6 | 1961 | |||
AL834230.1 | - | 5 | 1342 | |||
BC065530.1 | - | 11 | 3063 | 6152748 | ||
CR627478.1 | - | 4 | 642 |