Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBGCP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102610.1 | NP_001096080.1 | 7 | 778 | ||
NM_052903.4 | NP_443135.3 | 7 | 778 | |||
XM_005272540.2 | XP_005272597.1 | 7 | 685 | |||
XM_011543868.1 | XP_011542170.1 | 7 | 685 | |||
XM_011543869.2 | XP_011542171.1 | 7 | 682 | |||
XM_011543870.1 | XP_011542172.1 | 7 | 685 | |||
XM_017021890.1 | XP_016877379.1 | 7 | 685 | |||
XM_017021891.1 | XP_016877380.1 | 7 | 682 | |||
XM_017021892.1 | XP_016877381.1 | 7 | 682 | |||
XM_017021893.1 | XP_016877382.1 | 7 | 685 | |||
XM_017021894.1 | XP_016877383.1 | 4 | 333 | |||
XM_017021895.1 | XP_016877384.1 | 7 | 702 | |||
XM_017021898.1 | XP_016877387.1 | 2 | 92 | |||
XM_017021899.1 | XP_016877388.1 | 7 | 677 | |||
XM_017021900.1 | XP_016877389.1 | 7 | 646 | |||
GenBank mRNA | AB067486.2 | - | 7 | 692 | ||
AF272884.1 | - | 7 | 702 | |||
AK056416.1 | - | 7 | 677 | |||
AK289859.1 | - | 7 | 677 | |||
AK310897.1 | - | 7 | 674 | |||
BC071560.1 | - | 7 | 674 | 5297149 |