Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052905.3 | NP_443137.2 | 2 | 523 | ||
XM_005246263.2 | XP_005246320.1 | 2 | 590 | |||
XM_005246265.3 | XP_005246322.1 | 2 | 590 | |||
XM_011510530.1 | XP_011508832.1 | 2 | 590 | |||
XM_011510531.1 | XP_011508833.1 | 2 | 590 | |||
XM_011510532.2 | XP_011508834.1 | 2 | 590 | |||
XM_011510533.2 | XP_011508835.1 | 2 | 590 | |||
XM_011510534.1 | XP_011508836.1 | 2 | 590 | |||
XM_011510535.2 | XP_011508837.1 | 2 | 590 | |||
GenBank mRNA | AB067489.1 | - | 2 | 291 | ||
BC167159.1 | - | 2 | 291 | 8862581 | ||
CD105119.1 | - | 2 | 145 |