Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052911.2 | NP_443143.2 | 4 | 1975 | ||
XM_011525798.1 | XP_011524100.1 | 4 | 2105 | |||
XM_011525799.2 | XP_011524101.1 | 4 | 2238 | |||
GenBank mRNA | AB067498.1 | - | 4 | 1911 | ||
AK096746.1 | - | 4 | 1977 | |||
AL832041.1 | - | 4 | 2127 | |||
AL834200.1 | - | 1 | 683 | |||
BC089426.1 | - | 1 | 1395 | 3090307 |