Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM200A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258276.1 | NP_001245205.1 | 2 | 1570 | ||
NM_001258277.1 | NP_001245206.1 | 3 | 1634 | |||
NM_001258278.1 | NP_001245207.1 | 2 | 1589 | |||
NM_052913.2 | NP_443145.1 | 2 | 3688 | |||
XM_005266816.2 | XP_005266873.1 | 4 | 1713 | |||
XM_011535419.2 | XP_011533721.1 | 2 | 1390 | |||
XM_017010230.1 | XP_016865719.1 | 2 | 2365 | |||
GenBank mRNA | AB052099.1 | - | 2 | 3688 | ||
AB067500.1 | - | 3 | 2181 | |||
AK290166.1 | - | 2 | 1583 | |||
AK294359.1 | - | 1 | 1013 | |||
AK315824.1 | - | 2 | 1466 | |||
BC044246.1 | - | 3 | 1453 | 5760073 |