Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242508.1 | NP_001229437.1 | 10 | 1122 | ||
NM_018030.4 | NP_060500.3 | 8 | 1491 | |||
NM_080597.3 | NP_542164.2 | 22 | 2378 | |||
XM_006722380.3 | XP_006722443.1 | 21 | 2291 | |||
XM_006722382.3 | XP_006722445.1 | 21 | 2256 | |||
XM_017025530.1 | XP_016881019.1 | 22 | 2670 | |||
XM_017025531.1 | XP_016881020.1 | 20 | 1980 | |||
XM_017025533.1 | XP_016881022.1 | 8 | 974 | |||
XM_017025534.1 | XP_016881023.1 | 8 | 3502 | |||
GenBank mRNA | AF274714.1 | - | 8 | 1491 | ||
AF323726.2 | - | 22 | 2266 | |||
AF392449.1 | - | 8 | 1112 | |||
AF392450.1 | - | 22 | 2335 | |||
AK096358.1 | - | 22 | 2378 | |||
AK292895.1 | - | 23 | 2310 | |||
AK301862.1 | - | 10 | 1122 | |||
BC073854.1 | - | 6 | 698 | |||
BC098384.1 | - | 8 | 1170 | |||
BC136452.1 | - | 22 | 2275 | 9020441 | ||
BC144117.1 | - | 22 | 2275 | 9052637 |