Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201480.1 | NP_001188409.1 | 14, 15 | 1900 | ||
NM_001201481.1 | NP_001188410.1 | 12, 13 | 1732 | |||
NM_032523.3 | NP_115912.1 | 13, 14 | 1825 | |||
NM_145739.2 | NP_665682.1 | 12, 13 | 1413 | |||
XM_011510539.1 | XP_011508841.1 | 13, 14 | 1803 | |||
XM_017003265.1 | XP_016858754.1 | 15, 16 | 1972 | |||
XM_017003266.1 | XP_016858755.1 | 15, 16 | 1972 | |||
XM_017003267.1 | XP_016858756.1 | 13, 14 | 1667 | |||
XM_017003270.1 | XP_016858759.1 | 13, 14 | 1886 | |||
XM_017003273.1 | XP_016858762.1 | 11, 12 | 1635 | |||
GenBank mRNA | AB208898.1 | - | 14, 15 | 1900 | ||
AF323728.1 | - | 13, 14 | 1659 | |||
AF392448.1 | - | 13, 14 | 1659 | |||
AK027600.1 | - | 6, 7 | 700 | |||
AK093902.1 | - | 12, 13 | 1415 |