Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017784.4 | NP_060254.2 | 4 | 1360 | ||
XM_005264843.3 | XP_005264900.1 | 4 | 1136 | |||
XM_011533326.2 | XP_011531628.1 | 4 | 513 | |||
XM_017005667.1 | XP_016861156.1 | 4 | 1026 | |||
XM_017005668.1 | XP_016861157.1 | 4 | 1003 | |||
XM_017005669.1 | XP_016861158.1 | 4 | 532 | |||
GenBank mRNA | AF346291.1 | - | 4 | 806 | ||
AF392451.1 | - | 5 | 1070 | |||
AK000370.1 | - | 2 | 236 | |||
AL833247.1 | - | 4 | 463 | |||
BC003168.1 | - | 4 | 764 | 2905981 |