Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031909.2 | NP_114115.2 | 2 | 1164 | ||
XM_017017165.1 | XP_016872654.1 | 3 | 2482 | |||
XM_017017166.1 | XP_016872655.1 | 3 | 2258 | |||
GenBank mRNA | AF329838.1 | - | 2 | 1121 | ||
BC035628.1 | - | 2 | 947 | 5199881 |