Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031910.3 | NP_114116.3 | 2, 3 | 360 | ||
NM_182486.1 | NP_872292.1 | 2, 3 | 360 | |||
XM_011529857.1 | XP_011528159.1 | 3, 4 | 3217 | |||
XM_017028569.1 | XP_016884058.1 | 2, 3 | 2509 | |||
XM_017028570.1 | XP_016884059.1 | 4, 5 | 1127 | |||
XM_017028571.1 | XP_016884060.1 | 4, 5 | 2865 | |||
XM_017028572.1 | XP_016884061.1 | 4, 5 | 2854 | |||
XM_017028573.1 | XP_016884062.1 | 4, 5 | 643 | |||
XM_017028574.1 | XP_016884063.1 | 3, 4 | 436 | |||
GenBank mRNA | AF329842.1 | - | 2, 3 | 298 | ||
AK226149.1 | - | 1, 2 | 883 | |||
AY358374.1 | - | 4, 5 | 497 | |||
BC020551.1 | - | 2, 3 | 359 | 2900683 | ||
BU147265.1 | - | 2, 3 | 362 | |||
CB961508.1 | - | 2, 3 | 344 | |||
CR456451.1 | - | 2, 3 | 310 |