Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135674.1 | NP_001129146.1 | 2 | 839 | ||
NM_001135675.1 | NP_001129147.1 | 2 | 447 | |||
NM_001135676.1 | NP_001129148.1 | 2 | 132 | |||
NM_138436.3 | NP_612445.2 | 2 | 218 | |||
XM_005273398.4 | XP_005273455.1 | 2 | 753 | |||
GenBank mRNA | AK311932.1 | - | 2 | 193 | ||
BC013035.2 | - | 2 | 180 | 3535608 | ||
BI335341.1 | - | 2 | 630 | |||
BI463786.1 | - | 2 | 137 | |||
BI549696.1 | - | 2 | 452 | |||
DA702519.1 | - | 2 | 218 |